Sodium linoleate ≥95.0%

Supplier: TCI America

Synonyms: Linoleic Acid Sodium Salt

L0056-25G L0056-5G
TCL0056-025GEA 658.78 CAD
TCL0056-025G TCL0056-5G
Sodium linoleate ≥95.0%
Sodium linoleate

CAS Number: 822-17-3
MDL Number: MFCD00063202
Molecular Formula: C18H32O2
Molecular Weight: 302.43
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 192
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Linoleic Acid Sodium Salt

Formula: C₁₈H₃₂O₂
MW: 302.43 g/mol
Melting Pt: 192 °C
MDL Number: MFCD00063202
CAS Number: 822-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90e78dfc89a0d3e94cbdb0493d013e8cee956e0b&vid=90e78dfc89a0d3e94cbdb0493d013e8cee956e0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR