α-Linolenic acid methyl ester ≥70.0%
Supplier: TCI America
Synonyms:
Methyl cis,cis,cis-9,12,15-octadecatrienoate, Methyl linolenate, 9Z,12Z,15Z-Octadecatrienoic acid methyl ester, Methyl Linolenate, Linolenic acid methyl ester, cis,cis,cis-9,12,15-Octadecatrienoic acid methyl ester, Methyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, Methyl α-linolenate
CAS Number: 301-00-8
MDL Number: MFCD00135851
Molecular Formula: C19H32O2
Molecular Weight: 292.46
Purity/Analysis Method: >70.0% (GC)
Form: Clear Liquid
Boiling point (°C): 347
Flash Point (°C): 113
Specific Gravity (20/20): 0.90
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Linolenic Acid Methyl Ester
Formula:
C₁₉H₃₂O₂ MW: 292.46 g/mol Boiling Pt: 347 °C Melting Pt: –57 °C Density: 0.90 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00135851 CAS Number: 301-00-8 EINECS: 206-102-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...