Linalyl acetate ≥95.0%

Supplier: TCI America

Synonyms: 3,7-Dimethylocta-1,6-dien-3-yl acetate

L0049-25ML L0049-500ML
TCL0049-025MLEA 35.38 CAD
TCL0049-025ML TCL0049-500ML
Linalyl acetate ≥95.0%
Linalyl acetate

CAS Number: 115-95-7
MDL Number: MFCD00008907
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 85
Specific Gravity (20/20): 0.90

Formula: C₁₂H₂₀O₂
Boiling Pt: 220 °C
Density: 0.90 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00008907
CAS Number: 115-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f1a36232ac5f2f3436f86705d6965012557a76&vid=53f1a36232ac5f2f3436f86705d6965012557a76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR