Lignin (Dealkaline)

Supplier: TCI America

L0045-500G L0045-25G
TCL0045-500GEA 162.65 CAD
TCL0045-500G TCL0045-025G
Lignin (Dealkaline)
Lignin (dealkaline)

CAS Number: 9005-53-2
MDL Number: MFCD00147441
Form: Crystal
Color: Deep Yellow


Product Categories: Materials Chemistry, Polymers, Bioscience, Phenylpropanoids

MDL Number: MFCD00147441
CAS Number: 9005-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR