DL-Leucyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
L0038-1G
L0038-100MG
TCL0038-001GEA
195.92
CAD
TCL0038-001G
TCL0038-100MG
DL-Leucyl-DL-phenylalanine ≥98.0% (by titrimetric analysis)
DL-Leucyl-DL-phenylalanine
CAS Number: 56217-82-4
MDL Number: MFCD00037258
Molecular Formula: C15H22N2O3
Molecular Weight: 278.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Leu-DL-Phe-OH
Formula:
C₁₅H₂₂N₂O₃ |
MDL Number:
MFCD00037258 CAS Number: 56217-82-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...