DL-Leucylglycylglycine

Supplier: TCI America

L0033-1G L0033-100MG
TCL0033-001GEA 128.93 CAD
TCL0033-001G TCL0033-100MG
DL-Leucylglycylglycine
Proteins and Peptides

CAS Number: 4337-37-5 MDL Number: MFCD00064990 Molecular Formula: C10H19N3O4 Molecular Weight: 245.28 Purity/Analysis Method: 99.0% (T) Form: Crystal


  • Product Categories: Bioscience, Oligopeptides
  • Other Product Names / Synonyms: H-DL-Leu-Gly-Gly-OH
  • EINECS: 224-384-6


Product Categories: Bioscience, Oligopeptides Other Product Names / Synonyms: H-DL-Leu-Gly-Gly-OH

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5427269f830f9e1d7f1eb8b6dc775e6727fd3076&vid=5427269f830f9e1d7f1eb8b6dc775e6727fd3076&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR