DL-Leucylglycine 98,0

Supplier: TCI America

Synonyms: H-DL-Leu-Gly-OH, DL-Leucylglycine, 2-(2-Amino-4-methylpentanamido)acetic acid

L0030-1G L0030-100MG
TCL0030-001GEA 165.9 CAD
TCL0030-001G TCL0030-100MG
DL-Leucylglycine 98,0
DL-Leucylglycine

CAS Number: 615-82-7
MDL Number: MFCD00021729
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Leu-Gly-OH

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
MDL Number: MFCD00021729
CAS Number: 615-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91102da95497c3ca6ec07cca0ff1a3703d87055b&vid=91102da95497c3ca6ec07cca0ff1a3703d87055b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR