L-Leucic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-2-Hydroxy-4-methylpentanoic acid, L-alpha-Hydroxyisocaproic acid, (S)-2-Hydroxy-4-methylvaleric Acid
CAS Number: 13748-90-8
MDL Number: MFCD00064214
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 81
Specific rotation [a]20/D: -26 deg (C=2, NaOH sol.(1%))
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-Hydroxy-4-methylvaleric Acid
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Melting Pt: 81 °C |
MDL Number:
MFCD00064214 CAS Number: 13748-90-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...