Lauric anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

L0017-25G L0017-500G L0017-100G
TCL0017-025GEA 38.58 CAD
TCL0017-025G TCL0017-500G TCL0017-100G
Lauric anhydride ≥98.0% (by titrimetric analysis)
Lauric anhydride

CAS Number: 645-66-9
MDL Number: MFCD00008965
Molecular Formula: C24H46O3
Molecular Weight: 382.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 42
Flash Point (°C): 110

Formula: C₂₄H₄₆O₃
Melting Pt: 42 °C
Flash Pt: 110 °C
MDL Number: MFCD00008965
CAS Number: 645-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abca870c1eb5dffcad7358d86313a82c9b130d6e&vid=abca870c1eb5dffcad7358d86313a82c9b130d6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR