Amyl laurate ≥97.0%

Supplier: TCI America

L0012-500ML L0012-25ML
TCL0012-500MLEA 1051.33 CAD
TCL0012-500ML TCL0012-025ML
Amyl laurate ≥97.0%
Amyl laurate

CAS Number: 5350-03-8
MDL Number: MFCD00059218
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester

Formula: C₁₇H₃₄O₂
Density: 0.86 (20 °C)
MDL Number: MFCD00059218
CAS Number: 5350-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f48e8207490e9223de8ac4b2ff1dc542fd107a6c&vid=f48e8207490e9223de8ac4b2ff1dc542fd107a6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR