1,3-Adamantanediol ≥98.0%
Supplier: TCI America
A1531-5G
A1531-25G
TCA1531-005GEA
44.24
CAD
TCA1531-005G
TCA1531-025G
1,3-Adamantanediol ≥98.0%
1,3-Adamantanediol
CAS Number: 5001-18-3
MDL Number: MFCD00154013
Molecular Formula: C10H16O2
Molecular Weight: 168.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 258
Flash Point (°C): 202
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Dihydroxyadamantane
Formula:
C₁₀H₁₆O₂ Melting Pt: 258 °C Flash Pt: 202 °C |
MDL Number:
MFCD00154013 CAS Number: 5001-18-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...