Butyl (±)-lactate ≥98.0%

Supplier: TCI America

L0002-25G L0002-500G
TCL0002-025GEA 48.98 CAD
TCL0002-025G TCL0002-500G
Butyl (±)-lactate ≥98.0%
Butyl (±)-lactate

CAS Number: 138-22-7
MDL Number: MFCD00004519
Molecular Formula: C7H14O3
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 91
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Butyl 2-Hydroxypropionate, Lactic Acid Butyl Ester

Formula: C₇H₁₄O₃
Boiling Pt: 187 °C
Melting Pt: –28 °C
Density: 0.98 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00004519
CAS Number: 138-22-7
EINECS: 205-316-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4ea558deafd1f82bf657a9f6872a9216084ffaf&vid=a4ea558deafd1f82bf657a9f6872a9216084ffaf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR