DL-Lactamide 98,0 for synthesis

Supplier: TCI America

Synonyms: 2-Hydroxypropionamide, Lactamide

L0001-10G L0001-25G
TCL0001-010GEA 154.3 CAD
TCL0001-010G TCL0001-025G
DL-Lactamide 98,0 for synthesis
DL-Lactamide

CAS Number: 2043-43-8
MDL Number: MFCD00008017
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 2-Hydroxypropionamide

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00008017
CAS Number: 2043-43-8
EINECS: 218-049-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37db6a5e46ae205068107fa23b834bfd0e578023&vid=37db6a5e46ae205068107fa23b834bfd0e578023&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR