Calcium-4-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
K0023-25G
K0023-5G
TCK0023-025GEA
273.34
CAD
TCK0023-025G
TCK0023-005G
Calcium-4-methyl-2-oxovalerate hydrate ≥98.0% (by titrimetric analysis)
Calcium-4-methyl-2-oxovalerate hydrate
CAS Number: 51828-95-6
MDL Number: MFCD00004163
Molecular Formula: C6H10O3
Molecular Weight: 298.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 4-Methyl-2-oxovaleric Acid Calcium Salt
Formula:
C₆H₁₀O₃ |
MDL Number:
MFCD00004163 CAS Number: 51828-95-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...