Calcium-3-methyl-2-oxovalerate hydrate ≥98.0%
Supplier: TCI America
Synonyms:
3-Methyl-2-oxovaleric Acid Calcium Salt
K0022-25G
K0022-5G
TCK0022-025GEA
666.76
CAD
TCK0022-025G
TCK0022-005G
Calcium-3-methyl-2-oxovalerate hydrate ≥98.0%
Calcium-3-methyl-2-oxovalerate hydrate
CAS Number: 66872-75-1
MDL Number: MFCD00002583
Molecular Formula: C6H10O3
Molecular Weight: 298.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 3-Methyl-2-oxovaleric Acid Calcium Salt
Formula:
C₆H₁₀O₃ |
MDL Number:
MFCD00002583 CAS Number: 66872-75-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...