Kaempferol hydrate ≥97.0% (by HPLC)

Supplier: TCI America

K0018-100MG K0018-1G
TCK0018-100MGEA 290.88 CAD
TCK0018-100MG TCK0018-001G
Kaempferol hydrate ≥97.0% (by HPLC)
Kaempferol hydrate

CAS Number: 520-18-3
MDL Number: MFCD00016938
Molecular Formula: C15H10O6
Molecular Weight: 286.24
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,4',5,7-Tetrahydroxyflavone

Formula: C₁₅H₁₀O₆·H₂O
MDL Number: MFCD00016938
CAS Number: 520-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03ac9b6b809166959cc7992d983c7f424c4de580&vid=8835d976ff404ec7734f68c271bed147aa0c8c61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR