2-Oxovaleric acid ≥95.0%

Supplier: TCI America

K0015-1ML K0015-5ML
TCK0015-001MLEA 268.6 CAD
TCK0015-001ML TCK0015-005ML
2-Oxovaleric acid ≥95.0%
2-Oxovaleric acid

CAS Number: 1821-02-9
MDL Number: MFCD00066435
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Melting point (°C): 9
Flash Point (°C): 94
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Ketopentanoic Acid, 2-Oxopentanoic Acid, 2-Ketovaleric Acid

Formula: C₅H₈O₃
Boiling Pt: 179 °C
Melting Pt: 9 °C
Density: 1.12 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00066435
CAS Number: 1821-02-9

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR