5-Oxohexanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

K0014-25G K0014-5G
TCK0014-25GEA 1015.94 CAD
TCK0014-25G TCK0014-005G
5-Oxohexanoic acid ≥98.0% (by GC, titration analysis)
5-Oxohexanoic acid

CAS Number: 3128-06-1
MDL Number: MFCD00004412
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 160
Melting point (°C): 14
Flash Point (°C): 113
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 4-Acetylbutyric Acid, 5-Ketohexanoic Acid

Formula: C₆H₁₀O₃
Boiling Pt: 160 °C
Melting Pt: 14 °C
Density: 1.10 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004412
CAS Number: 3128-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9947a4a45f8adf724cb3075063fbb802503f5c17&vid=9947a4a45f8adf724cb3075063fbb802503f5c17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR