Dimethyl-2-oxoglutarate ≥95.0%

Supplier: TCI America

Synonyms: 2-Ketoglutaric Acid Dimethyl Ester, Dimethyl 2-Ketoglutarate, 2-Oxoglutaric Acid Dimethyl Ester

K0013-25ML K0013-5ML
TCK0013-025MLEA 426.6 CAD
TCK0013-025ML TCK0013-005ML
Dimethyl-2-oxoglutarate ≥95.0%
Dimethyl-2-oxoglutarate

CAS Number: 13192-04-6
MDL Number: MFCD00048052
Molecular Formula: C7H10O5
Molecular Weight: 174.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: Dimethyl 2-Ketoglutarate, 2-Ketoglutaric Acid Dimethyl Ester, 2-Oxoglutaric Acid Dimethyl Ester

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 117 °C
Density: 1.21 (20 °C)
MDL Number: MFCD00048052
CAS Number: 13192-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=931c9383723993b050602b63f8831111742ecdfe&vid=931c9383723993b050602b63f8831111742ecdfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR