α-Ketoglutaric acid disodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Disodium 2-Ketoglutate, Disodium 2-Oxoglutarate, 2-Ketoglutaric Acid Disodium Salt

K0007-5G K0007-25G
TCK0007-5GEA 47.4 CAD
TCK0007-5G TCK0007-025G
α-Ketoglutaric acid disodium salt ≥98.0% (by titrimetric analysis)
α-Ketoglutaric acid disodium salt

CAS Number: 305-72-6
MDL Number: MFCD00043347
Molecular Formula: C5H6O5
Molecular Weight: 190.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Disodium 2-Ketoglutarate, 2-Ketoglutaric Acid Disodium Salt, 2-Oxoglutaric Acid Disodium Salt

Formula: C₅H₄Na₂O₅
MW: 190.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00043347
CAS Number: 305-72-6
EINECS: 206-167-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a612a78589f3ec90bd0dd89b30e0972194e083d2&vid=05ecf6af4aa50ed3c75242d351b191db54e25cfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR