Jasmonic acid (mixture of isomers) ≥85.0%

Supplier: TCI America

J0004-5G J0004-1G
TCJ0004-5GEA 1132.86 CAD
TCJ0004-5G TCJ0004-1G
Jasmonic acid (mixture of isomers) ≥85.0%
Jasmonic acid (mixture of isomers)

CAS Number: 221682-41-3
Molecular Formula: C12H18O3
Molecular Weight: 210.27
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.07


Product Categories: Bioscience, Jasmonates
Other Product Names / Synonyms: 3-(Carboxymethyl)-2-(2-pentenyl)cyclopentanone, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid

Formula: C₁₂H₁₈O₃
Density: 1.07 (20 °C)
CAS Number: 221682-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=911586ca7e3faf77ecfe2d058dcda8b120bbb72a&vid=911586ca7e3faf77ecfe2d058dcda8b120bbb72a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR