(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0%

Supplier: TCI America

Synonyms: (4S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone, (S)-4-Isopropyl-5,5-diphenyloxazolidin-2-one

I0762-5G I0762-1G
TCI0762-5GEA 943.89 CAD
TCI0762-5G TCI0762-1G
(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0%
(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone

CAS Number: 184346-45-0
MDL Number: MFCD03093556
Molecular Formula: C18H19NO2
Molecular Weight: 281.36
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 254
Specific rotation [a]20/D: -246 deg (C=0.6, CH2Cl2)

Formula: C₁₈H₁₉NO₂
Melting Pt: 254 °C
MDL Number: MFCD03093556
CAS Number: 184346-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0721bc0031b77dc8a571fdc9573528877ac40a5&vid=c0721bc0031b77dc8a571fdc9573528877ac40a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR