(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0%

Supplier: TCI America

Synonyms: (R)-4-Isopropyl-5,5-diphenyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone

I0761-1G I0761-5G
TCI0761-1GEA 208.56 CAD
TCI0761-1G TCI0761-5G
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0%
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone

CAS Number: 191090-32-1
MDL Number: MFCD03093555
Molecular Formula: C18H19NO2
Molecular Weight: 281.36
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 254
Specific rotation [a]20/D: 247 deg (C=0.6, CH2Cl2)

Formula: C₁₈H₁₉NO₂
Melting Pt: 254 °C
MDL Number: MFCD03093555
CAS Number: 191090-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a9d124741ff957d60a013f347c59b866b1b7413&vid=8a9d124741ff957d60a013f347c59b866b1b7413&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR