Indole-6-carbaldehyd ≥98.0%

Supplier: TCI America

Synonyms: Indole-6-carboxaldehyde, 1H-Indole-6-carbaldehyde

I0743-1G I0743-5G
TCI0743-1GEA 162.74 CAD
TCI0743-1G TCI0743-5G
Indole-6-carbaldehyd ≥98.0%
Indole-6-carbaldehyd

CAS Number: 1196-70-9
MDL Number: MFCD02179596
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129


Other Product Names / Synonyms: 6-Formylindole

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 129 °C
MDL Number: MFCD02179596
CAS Number: 1196-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef03959955b74768c633b35143e88aa5e42fbbdc&vid=ef03959955b74768c633b35143e88aa5e42fbbdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR