Imidazo[1,2-b]pyridazine ≥98.0%
Supplier: TCI America
I0742-1G
I0742-5G
TCI0742-1GEA
33.76
CAD
TCI0742-1G
TCI0742-5G
Imidazo[1,2-b]pyridazine ≥98.0%
Imidazo[1,2-b]pyridazine
CAS Number: 766-55-2
MDL Number: MFCD07782103
Molecular Formula: C6H5N3
Molecular Weight: 119.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Lambda max.: 222 nm (MeOH)
Formula:
C₆H₅N₃ Melting Pt: 54 °C |
MDL Number:
MFCD07782103 CAS Number: 766-55-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...