L-Isoleucinamide hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
I0731-1G
I0731-5G
TCI0731-1GEA
41.08
CAD
TCI0731-1G
TCI0731-5G
L-Isoleucinamide hydrochloride ≥98.0% (by total nitrogen basis)
L-Isoleucinamide hydrochloride
CAS Number: 10466-56-5
MDL Number: MFCD00058476
Molecular Formula: C6H14N2O
Molecular Weight: 166.65
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 246
Specific rotation [a]20/D: 22 deg (C=1, H2O)
Formula:
C₆H₁₄N₂O Melting Pt: 246 °C |
MDL Number:
MFCD00058476 CAS Number: 10466-56-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...