2-Isopropyl-N,2,3-trimethylbutyramide ≥98.0%
Supplier: TCI America
I0729-25G
I0729-5G
TCI0729-25GEA
104.47
CAD
TCI0729-25G
TCI0729-5G
2-Isopropyl-N,2,3-trimethylbutyramide ≥98.0%
2-Isopropyl-N,2,3-trimethylbutyramide
CAS Number: 51115-67-4
MDL Number: MFCD00130070
Molecular Formula: C10H21NO
Molecular Weight: 171.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 64
Formula:
C₁₀H₂₁NO Boiling Pt: 85 °C Melting Pt: 64 °C |
MDL Number:
MFCD00130070 CAS Number: 51115-67-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...