1,2-O-Isopropylidene-ɑ-D-xylofuranose ≥98.0%

Supplier: TCI America

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

I0721-25G I0721-5G
TCI0721-25GEA 302.51 CAD
TCI0721-25G TCI0721-5G
1,2-O-Isopropylidene-ɑ-D-xylofuranose ≥98.0%
1,2-O-Isopropylidene-alpha-D-xylofuranose

CAS Number: 20031-21-4
MDL Number: MFCD00063295
Molecular Formula: C8H14O5
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70
Specific rotation [a]20/D: -19 deg (C=1, H2O)

Formula: C₈H₁₄O₅
MW: 190.20 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00063295
CAS Number: 20031-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14b4b1997bf6a54fa9c173f976a9f7ea62ddab72&vid=14b4b1997bf6a54fa9c173f976a9f7ea62ddab72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR