12-Aminododecan-1-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 12-Amino-1-dodecanol

A1524-1G A1524-5G
TCA1524-1GEA 325.64 CAD
TCA1524-1G TCA1524-5G
12-Aminododecan-1-ol ≥98.0% (by GC, titration analysis)
12-Aminododecan-1-ol

CAS Number: 67107-87-3
MDL Number: MFCD01851111
Molecular Formula: C12H27NO
Molecular Weight: 201.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81

Formula: C₁₂H₂₇NO
Melting Pt: 81 °C
MDL Number: MFCD01851111
CAS Number: 67107-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e20b9ac538644ae212eea3de33b633e9904b5bc8&vid=e20b9ac538644ae212eea3de33b633e9904b5bc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR