Irsogladine maleate ≥98.0%
Supplier: TCI America
Synonyms:
6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine maleate, 2,4-Diamino-6-(2,5-dichlorophenyl)-1,3,5-triazine Maleate
I0719-5G
I0719-1G
TCI0719-5GEA
608.3
CAD
TCI0719-5G
TCI0719-1G
Irsogladine maleate ≥98.0%
Irsogladine maleate
CAS Number: 84504-69-8
MDL Number: MFCD00873566
Molecular Formula: C9H7Cl2N5
Molecular Weight: 372.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2,4-Diamino-6-(2,5-dichlorophenyl)-1,3,5-triazine Maleate
Formula:
C₉H₇Cl₂N₅ MW: 372.16 g/mol |
MDL Number:
MFCD00873566 CAS Number: 84504-69-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...