N2-Isobutyryl-2'-deoxyguanosine 98,0
Supplier: TCI America
Synonyms:
N-(9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide
I0700-1G
I0700-100MG
TCI0700-1GEA
611.46
CAD
TCI0700-1G
TCI0700-100MG
N2-Isobutyryl-2'-deoxyguanosine 98,0
N2-Isobutyryl-2'-deoxyguanosine
CAS Number: 68892-42-2
MDL Number: MFCD00010060
Molecular Formula: C14H19N5O5
Molecular Weight: 337.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 300
Storage Temperature: <0°C
Formula:
C₁₄H₁₉N₅O₅ MW: 337.34 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00010060 CAS Number: 68892-42-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...