N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine ≥99.0% (by HPLC)

Supplier: TCI America

I0697-1G I0697-5G
TCI0697-1GEA 54.51 CAD
TCI0697-1G TCI0697-5G
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine ≥99.0% (by HPLC)
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine

CAS Number: 68892-41-1
MDL Number: MFCD00010059
Molecular Formula: C35H37N5O7
Molecular Weight: 639.71
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 155


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: iBu-DMT-dG, 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine

Formula: C₃₅H₃₇N₅O₇
Melting Pt: 155 °C
MDL Number: MFCD00010059
CAS Number: 68892-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b89f89b245e39c29aa0c5c4f622264998d181e4&vid=5b89f89b245e39c29aa0c5c4f622264998d181e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR