4-Iodophenylsulfur pentafluoride ≥94.0%

Supplier: TCI America

I0695-1G I0695-5G
TCI0695-1GEA 308.1 CAD
TCI0695-1G TCI0695-5G
4-Iodophenylsulfur pentafluoride ≥94.0%
4-Iodophenylsulphur pentafluoride

CAS Number: 286947-68-0
MDL Number: MFCD03788514
Molecular Formula: C6H4F5IS
Molecular Weight: 330.05
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Boiling point (°C): 52
Melting point (°C): 36
Storage Temperature: 0-10°C

Formula: C₆H₄F₅IS
MW: 330.600 g/mol
Boiling Pt: 52 °C
Melting Pt: 36 °C
MDL Number: MFCD03788514
CAS Number: 286947-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40b96c6741213a0946591200aa4081d5574d1e8&vid=d40b96c6741213a0946591200aa4081d5574d1e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR