10-Aminodecan-1-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1523-1G A1523-5G
TCA1523-1GEA 284.01 CAD
TCA1523-1G TCA1523-5G
10-Aminodecan-1-ol ≥98.0% (by GC, titration analysis)
10-Aminodecan-1-ol

CAS Number: 23160-46-5
MDL Number: MFCD02094313
Molecular Formula: C10H23NO
Molecular Weight: 173.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 72

Formula: C₁₀H₂₃NO
Boiling Pt: 154 °C
Melting Pt: 72 °C
MDL Number: MFCD02094313
CAS Number: 23160-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d2708d2eea3eaa9d80b736463abeddb638fb30&vid=26d2708d2eea3eaa9d80b736463abeddb638fb30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR