L-Isoleucine-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
I0685-5G
I0685-1G
TCI0685-5GEA
126.4
CAD
TCI0685-5G
TCI0685-1G
L-Isoleucine-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Isoleucine-tert-butyl ester hydrochloride
CAS Number: 69320-89-4
MDL Number: MFCD00058015
Molecular Formula: C10H21NO2
Molecular Weight: 223.74
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 160
Specific rotation [a]20/D: 15 deg (C=1, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Ile-OtBu.HCl
Formula:
C₁₀H₂₁NO₂ Melting Pt: 160 °C |
MDL Number:
MFCD00058015 CAS Number: 69320-89-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...