2-(1H-Imidazol-1-yl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Imidazoleacetic acid, Imidazol-1-yl-acetic acid, 1-(Carboxymethyl)imidazole, 2-(1-Imidazolyl)acetic acid

I0670-25G I0670-5G
TCI0670-25GEA 143.78 CAD
TCI0670-25G TCI0670-5G
2-(1H-Imidazol-1-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(1H-Imidazol-1-yl)acetic acid

CAS Number: 22884-10-2
MDL Number: MFCD01721800
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 269


Other Product Names / Synonyms: 2-(1-Imidazolyl)acetic Acid, 1-(Carboxymethyl)imidazole

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
Melting Pt: 269 °C
MDL Number: MFCD01721800
CAS Number: 22884-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ee72d543cc17bbb718afad121671e2bacf2ebbf&vid=6ee72d543cc17bbb718afad121671e2bacf2ebbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR