Indole-3-acetamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

I0668-5G I0668-25G
TCI0668-5GEA 71.1 CAD
TCI0668-5G TCI0668-25G
Indole-3-acetamide ≥98.0% (by HPLC, total nitrogen)
Indole-3-acetamide

CAS Number: 879-37-8
MDL Number: MFCD00005641
Molecular Formula: C10H10N2O
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 153

Formula: C₁₀H₁₀N₂O
Melting Pt: 153 °C
MDL Number: MFCD00005641
CAS Number: 879-37-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eb6a402fd11e9033fe7314338602af814add9d3&vid=6eb6a402fd11e9033fe7314338602af814add9d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR