3-Isoamyl-6-methyl-2-heptyl myristate ≥95.0%

Supplier: TCI America

Synonyms: Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate

I0662-25G I0662-500G
TCI0662-25GEA 42.66 CAD
TCI0662-25G TCI0662-500G
3-Isoamyl-6-methyl-2-heptyl myristate ≥95.0%
3-Isoamyl-6-methyl-2-heptyl myristate

CAS Number: 88332-30-3
Molecular Formula: C27H54O2
Molecular Weight: 410.73
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 205
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester

Formula: C₂₇H₅₄O₂
MW: 410.73 g/mol
Density: 0.85 (20 °C)
Flash Pt: 205 °C
CAS Number: 88332-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41d5e158d14ce521edc23b465570945d64e99d26&vid=41d5e158d14ce521edc23b465570945d64e99d26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR