8-Aminooctan-1-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1522-1G A1522-5G
TCA1522-1GEA 213.3 CAD
TCA1522-1G TCA1522-5G
8-Aminooctan-1-ol ≥98.0% (by GC, titration analysis)
8-Aminooctan-1-ol

CAS Number: 19008-71-0
MDL Number: MFCD02094315
Molecular Formula: C8H19NO
Molecular Weight: 145.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 65

Formula: C₈H₁₉NO
Melting Pt: 65 °C
MDL Number: MFCD02094315
CAS Number: 19008-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc3328cede51729758c4b1a4b244fbab469dff2f&vid=cc3328cede51729758c4b1a4b244fbab469dff2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR