(Isocyanoimino)triphenylphosphorane ≥95.0% (by HPLC)
Supplier: TCI America
I0643-25G
I0643-5G
TCI0643-25GEA
996.66
CAD
TCI0643-25G
TCI0643-5G
(Isocyanoimino)triphenylphosphorane ≥95.0% (by HPLC)
(Isocyanoimino)triphenylphosphorane
CAS Number: 73789-56-7
Molecular Formula: C19H15N2P
Molecular Weight: 302.32
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 152
Formula:
C₁₉H₁₅N₂P MW: 302.32 g/mol Melting Pt: 152 °C |
MDL Number:
MFCD09038528 CAS Number: 73789-56-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...