4-Chloro-6-methoxypyrimidin-2-amine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
A1520-5G
A1520-25G
TCA1520-005GEA
34.76
CAD
TCA1520-005G
TCA1520-025G
4-Chloro-6-methoxypyrimidin-2-amine ≥95.0% (by HPLC, titration analysis)
4-Chloro-6-methoxypyrimidin-2-amine
CAS Number: 5734-64-5
MDL Number: MFCD00233535
Molecular Formula: C5H6ClN3O
Molecular Weight: 159.57
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 170
Formula:
C₅H₆ClN₃O Melting Pt: 170 °C |
MDL Number:
MFCD00233535 CAS Number: 5734-64-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...