Isobornyl methacrylate ≥85.0% stabilized

Supplier: TCI America

I0617-25G I0617-500G
TCI0617-25GEA 49.82 CAD
TCI0617-25G TCI0617-500G
Isobornyl methacrylate ≥85.0% stabilized
Isobornyl methacrylate

(stabilized with MEHQ)
CAS Number: 7534-94-3
MDL Number: MFCD00081070
Molecular Formula: C14H22O2
Molecular Weight: 222.33
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 144
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents, Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: Methacrylic Acid Isobornyl Ester

Formula: C₁₄H₂₂O₂
Boiling Pt: 129 °C
Density: 0.98 (20 °C)
Flash Pt: 144 °C
MDL Number: MFCD00081070
CAS Number: 7534-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4f6ef1af813237de268811d3e4b3f99e114c8c3&vid=b4f6ef1af813237de268811d3e4b3f99e114c8c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR