1H-Imidazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Imidazole-4-carboxylic acid

I0607-1G I0607-5G
TCI0607-1GEA 37.92 CAD
TCI0607-1G TCI0607-5G
1H-Imidazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1H-Imidazole-4-carboxylic acid

CAS Number: 1072-84-0
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 284
Storage Temperature: 0-10°C

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 284 °C
MDL Number: MFCD00082203
CAS Number: 1072-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebabd49bda6c849fc4d9f41d291b5a8ba8061b51&vid=ebabd49bda6c849fc4d9f41d291b5a8ba8061b51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR