Ethyl Indoline-2-carboxylate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl indoline-2-carboxylate, Indoline-2-carboxylic Acid Ethyl Ester

I0595-25G I0595-5G
TCI0595-25GEA 1847.02 CAD
TCI0595-25G TCI0595-5G
Ethyl Indoline-2-carboxylate ≥95.0%
Ethyl Indoline-2-carboxylate

CAS Number: 50501-07-0
MDL Number: MFCD06797114
Molecular Formula: C11H13NO2
Molecular Weight: 191.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 55
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Indoline-2-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
Melting Pt: 55 °C
MDL Number: MFCD06797114
CAS Number: 50501-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d316e9e306d02a80fb18a241037f7a7fe5cfbae4&vid=d316e9e306d02a80fb18a241037f7a7fe5cfbae4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR