5-Indoleboronic acid pinacol ester ≥98.0%
Supplier: TCI America
Synonyms:
Indole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
I0590-1G
I0590-5G
TCI0590-1GEA
134.3
CAD
TCI0590-1G
TCI0590-5G
5-Indoleboronic acid pinacol ester ≥98.0%
5-Indoleboronic acid pinacol ester
CAS Number: 269410-24-4
MDL Number: MFCD03789263
Molecular Formula: C14H18BNO2
Molecular Weight: 243.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 5-Indoleboronic Acid Pinacol Ester
Formula:
C₁₄H₁₈BNO₂ MW: 243.1092 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD03789263 CAS Number: 269410-24-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...