4-(2-Aminoethyl)-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1517-5G
A1517-25G
TCA1517-005GEA
309.05
CAD
TCA1517-005G
TCA1517-025G
4-(2-Aminoethyl)-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
4-(2-Aminoethyl)-1-benzylpiperidine
CAS Number: 86945-25-7
MDL Number: MFCD02093423
Molecular Formula: C14H22N2
Molecular Weight: 218.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 2-(1-Benzyl-4-piperidyl)ethylamine
Formula:
C₁₄H₂₂N₂ Density: 0.99 (20 °C) |
MDL Number:
MFCD02093423 CAS Number: 86945-25-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...