Bis(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0584-500G I0584-25G
TCI0584-500GEA 489.33 CAD
TCI0584-500G TCI0584-25G
Bis(2-carboxyethyl)isocyanurate ≥98.0% (by titrimetric analysis)
Bis(2-carboxyethyl)isocyanurate

CAS Number: 2904-40-7
MDL Number: MFCD04038422
Molecular Formula: C9H11N3O7
Molecular Weight: 273.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 289


Other Product Names / Synonyms: Isocyanuric Acid Bis(2-carboxyethyl) Ester

Formula: C₉H₁₁N₃O₇
Melting Pt: 289 °C
MDL Number: MFCD04038422
CAS Number: 2904-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d558325768c9bab382c4c2f581b211c568d420ea&vid=d558325768c9bab382c4c2f581b211c568d420ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR