(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥95.0%
Supplier: TCI America
I0582-1G
I0582-250MG
TCI0582-1GEA
406.06
CAD
TCI0582-1G
TCI0582-250MG
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥95.0%
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)
CAS Number: 131457-46-0
MDL Number: MFCD00192245
Molecular Formula: C21H22N2O2
Molecular Weight: 334.42
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific rotation [a]20/D: -155 deg (C=1, EtOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (S,S)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)
Formula:
C₂₁H₂₂N₂O₂ Flash Pt: 110 °C |
MDL Number:
MFCD00192245 CAS Number: 131457-46-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...