2-Isopropyl-3-methoxypyrazine ≥98.0%

Supplier: TCI America

I0577-1G I0577-5G
TCI0577-1GEA 101.12 CAD
TCI0577-1G TCI0577-5G
2-Isopropyl-3-methoxypyrazine ≥98.0%
2-Isopropyl-3-methoxypyrazine

CAS Number: 25773-40-4
MDL Number: MFCD00006135
Molecular Formula: C8H12N2O
Molecular Weight: 152.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 67

Formula: C₈H₁₂N₂O
Flash Pt: 67 °C
MDL Number: MFCD00006135
CAS Number: 25773-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5902236f8d833581347bbab6b8623287e9215bbf&vid=5902236f8d833581347bbab6b8623287e9215bbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR