(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) ≥97.0%

Supplier: TCI America

Synonyms: (S,S)-(-)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline), (4S,4'S)-2,2'-(Propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole), (S,S)-(-)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane, 2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline]

I0567-100MG I0567-1G
TCI0567-100MGEA 139.04 CAD
TCI0567-100MG TCI0567-001G
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) ≥97.0%
(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline)

CAS Number: 131833-93-7
MDL Number: MFCD00192243
Molecular Formula: C17H30N2O2
Molecular Weight: 294.44
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: -118 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-(-)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane, (S,S)-(-)-2,2'-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline)

Formula: C₁₇H₃₀N₂O₂
Melting Pt: 90 °C
MDL Number: MFCD00192243
CAS Number: 131833-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02df44c856c58755fcca31d031484a83d352a617&vid=02df44c856c58755fcca31d031484a83d352a617&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR